MilliporeSigma
  • Towards a method for loudness-based analysis of the sound of one's own voice.
Towards a method for loudness-based analysis of the sound of one's own voice.
Logopedics, phoniatrics, vocology (2013-04-11)
Manuj Yadav, Densil Cabrera, Dianna T Kenny
ABSTRACT
This paper outlines the steps in objectively estimating the time-varying loudness of one's own voice in a room (i.e. autophonic loudness). Voice recordings, made with a near-mouth microphone, are converted to the sound that reaches the two eardrums of the talking (or singing)-listener by convolving them with the impulse responses from the mouth to the respective ears of an anthropomorphic head and torso. The influences of bone-conducted sound and room reflections are taken into account. These convolved recordings are then processed with a computational time-varying loudness model. The method is demonstrated by a short case study, and the results illustrate something of the benefit of loudness analysis over sound pressure level analysis for representing autophonic loudness.
Product Description
Supelco
VOCOL® Capillary GC Column, L × I.D. 105 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 1.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.80 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.50 μm