MilliporeSigma
  • Role of μ-opioid receptor in parafascicular nucleus of thalamus on morphine-induced antinociception in a rat model of acute trigeminal pain.
Role of μ-opioid receptor in parafascicular nucleus of thalamus on morphine-induced antinociception in a rat model of acute trigeminal pain.
Veterinary research forum : an international quarterly journal (2017-05-06)
Esmaeal Tamaddonfard, Amir Erfanparast
ABSTRACT
The parafascicular nucleus (PFN) of thalamus, as a supraspinal structure, has an important role in processing of nociceptive information. In addition, μ-opioid receptor contributes to supraspinal modulation of nociception. In the present study, the effects of microinjection of naloxone (a non-specific opioid-receptor antagonist) and naloxonazine (a specific μ-opioid receptor antagonist) were investigated on morphine-induced antinociception in a rat model of acute trigeminal pain. Right and left sides of PFN of thalamus were implanted with two guide cannulas. Acute trigeminal pain was induced by local corneal surface application of hypertonic saline and the number of eye wipes as a pain index was recorded for 30 sec. Microinjection of morphine at doses of 1, 2 and 4 μg per site significantly (