MilliporeSigma
Phosphorylation-induced conformation of β
Nature communications (2018-01-18)
Yutaro Shiraishi, Mei Natsume, Yutaka Kofuku, Shunsuke Imai, Kunio Nakata, Toshimi Mizukoshi, Takumi Ueda, Hideo Iwaï, Ichio Shimada
ABSTRACT
The C-terminal region of G-protein-coupled receptors (GPCRs), stimulated by agonist binding, is phosphorylated by GPCR kinases, and the phosphorylated GPCRs bind to arrestin, leading to the cellular responses. To understand the mechanism underlying the formation of the phosphorylated GPCR-arrestin complex, we performed NMR analyses of the phosphorylated β