MilliporeSigma
  • Controlled growth and optical response of a semi-hollow plasmonic nanocavity and ultrathin sulfide nanosheets on Au/Ag platelets.
Controlled growth and optical response of a semi-hollow plasmonic nanocavity and ultrathin sulfide nanosheets on Au/Ag platelets.
Nanoscale (2018-01-03)
Ying Xie, Gui-Ming Pan, Ying-Ying Li, Kai Chen, Yong-Jie Lin, Li Zhou, Qu-Quan Wang
ABSTRACT
Herein, we report a strategy to construct a semi-hollow plasmonic nanocavity and grow ultrathin sulfide nanosheets inside. The competition and cooperation of Au deposition with Ag etching based on flat Ag nanoplates are proposed. For the establishment of the semi-hollow nanocavity, Au shells are grown on Ag nanoplates, which serve as a stable frame, followed by partial etching of the Ag nanoplates. By controlling the thickness of the initial Ag nanoplates or the injected amount of etchant, the nanocavity size is fine-tuned. Significantly, the remaining unetched Ag layers provide a flat platform for the growth of 2D ultrathin sulfides of Ag
Sigma-Aldrich
Bismuth(III) acetate, ≥99.99% trace metals basis