MilliporeSigma
  • The Long Noncoding RNA TP73-AS1 Interacted with miR-124 to Modulate Glioma Growth by Targeting Inhibitor of Apoptosis-Stimulating Protein of p53.
The Long Noncoding RNA TP73-AS1 Interacted with miR-124 to Modulate Glioma Growth by Targeting Inhibitor of Apoptosis-Stimulating Protein of p53.
DNA and cell biology (2018-02-08)
Shuai Xiao, Rensheng Wang, Xiangwei Wu, Wen Liu, Shanshan Ma
ABSTRACT
P73 antisense RNA 1T (non-protein coding), known as TP73-AS1 or PDAM, is a long noncoding RNA (lncRNA), which may regulate apoptosis by regulation of p53-dependent antiapoptotic genes. An abnormal change of TP73-AS1 expression was noticed in cancers. The effects of TP73-AS1 in brain glioma growth and the underlying mechanism remain unclear so far. In this study, the effect of TP73-AS1 in human brain glioma cell lines and clinical tumor samples was detected so as to reveal its role and function. In this study, TP73-AS1 was specifically upregulated in brain glioma cell lines and promoted glioma cell growth through targeting miR-124. TP73-AS1 knocking down suppressed human brain glioma cell proliferation, invasion, and metastasis in vitro. The inhibitory effect of TP73-AS1 knocking down on glioma cell proliferation and invasion could partly be restored by miR-124 inhibition. In addition, miR-124-dependent inhibitor of apoptosis-stimulating protein of p53 (iASPP) regulation was required in TP73-AS1-induced brain glioma cell growth. Data from this study revealed that TP73-AS1 inhibited the brain glioma growth and metastasis as a competing endogenous RNA (ceRNA) through miR-124-dependent iASPP regulation. In conclusion, we regarded TP73-AS1 as an oncogenic lncRNA promoting brain glioma proliferation and metastasis and a potential target for human brain glioma treatment.
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA