MilliporeSigma
  • A general method for the synthesis of unsymmetrically substituted ureas via palladium-catalyzed amidation.
A general method for the synthesis of unsymmetrically substituted ureas via palladium-catalyzed amidation.
Organic letters (2009-01-31)
Brian J Kotecki, Dilinie P Fernando, Anthony R Haight, Kirill A Lukin
ABSTRACT
A general and practical method for the preparation of unsymmetrically substituted ureas has been developed utilizing palladium-catalyzed amidation. Both aryl bromides and chlorides, as well as heteroaryl chlorides, have been coupled to aryl, benzyl, and aliphatic ureas by using a novel nonproprietary bipyrazole ligand (bippyphos).
Sigma-Aldrich
5-(Di-tert-butylphosphino)-1′, 3′, 5′-triphenyl-1′H-[1,4′]bipyrazole, 97%