MilliporeSigma
  • First determination of fullerenes in the Austrian market and environment: quantitative analysis and assessment.
First determination of fullerenes in the Austrian market and environment: quantitative analysis and assessment.
Environmental science and pollution research international (2017-10-20)
Susanna Zakaria, Eleonore Fröhlich, Günter Fauler, Anna Gries, Stefan Weiß, Sigrid Scharf
ABSTRACT
This study forms the first report on analyzing fullerenes in the Austrian environment and cosmetic products available on the Austrian market. We developed, optimized, and validated a novel method for the analysis of C
Sigma-Aldrich
N-Methylfulleropyrrolidine, 99% (HPLC)