MilliporeSigma
  • UCHL1-HIF-1 axis-mediated antioxidant property of cancer cells as a therapeutic target for radiosensitization.
UCHL1-HIF-1 axis-mediated antioxidant property of cancer cells as a therapeutic target for radiosensitization.
Scientific reports (2017-08-02)
Ryota Nakashima, Yoko Goto, Sho Koyasu, Minoru Kobayashi, Akiyo Morinibu, Michio Yoshimura, Masahiro Hiraoka, Ester M Hammond, Hiroshi Harada
ABSTRACT
Hypoxia-inducible factor 1 (HIF-1) has been recognized as an important mediator of the reprogramming of carbohydrate metabolic pathways from oxidative phosphorylation to accelerated glycolysis. Although this reprogramming has been associated with the antioxidant and radioresistant properties of cancer cells, gene networks triggering the HIF-1-mediated reprogramming and molecular mechanisms linking the reprogramming with radioresistance remain to be determined. Here, we show that Ubiquitin C-terminal hydrolase-L1 (UCHL1), which we previously identified as a novel HIF-1 activator, increased the radioresistance of cancer cells by producing an antioxidant, reduced glutathione (GSH), through HIF-1-mediated metabolic reprogramming. A luciferase assay to monitor HIF-1 activity demonstrated that the overexpression of UCHL1, but not its deubiquitination activity-deficient mutant (UCHL1 C90S), upregulated HIF-1 activity by stabilizing the regulatory subunit of HIF-1 (HIF-1α) in a murine breast cancer cell line, EMT6. UCHL1 overexpression induced the reprogramming of carbohydrate metabolism and increased NADPH levels in a pentose phosphate pathway (PPP)-dependent manner. The UCHL1-mediated reprogramming elevated intracellular GSH levels, and consequently induced a radioresistant phenotype in a HIF-1-dependent manner. The pharmacological inhibition of PPP canceled the UCHL1-mediated radioresistance. These results collectively suggest that cancer cells acquire antioxidant and radioresistant phenotypes through UCHL1-HIF-1-mediated metabolic reprogramming including the activation of PPP and provide a rational basis for targeting this gene network for radiosensitization.
Sigma-Aldrich
Anti-UCHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution