MilliporeSigma
  • Expression of CD38 in myeloma bone niche: A rational basis for the use of anti-CD38 immunotherapy to inhibit osteoclast formation.
Expression of CD38 in myeloma bone niche: A rational basis for the use of anti-CD38 immunotherapy to inhibit osteoclast formation.
Oncotarget (2017-09-17)
Federica Costa, Denise Toscani, Antonella Chillemi, Valeria Quarona, Marina Bolzoni, Valentina Marchica, Rosanna Vescovini, Cristina Mancini, Eugenia Martella, Nicoletta Campanini, Chiara Schifano, Sabrina Bonomini, Fabrizio Accardi, Alberto L Horenstein, Franco Aversa, Fabio Malavasi, Nicola Giuliani
ABSTRACT
It is known that multiple myeloma (MM) cells express CD38 and that a recently developed human anti-CD38 monoclonal antibody Daratumumab mediates myeloma killing. However, the expression of CD38 and other functionally related ectoenzymes within the MM bone niche and the potential effects of Daratumumab on bone cells are still unknown. This study firstly defines by flow cytometry and immunohistochemistry the expression of CD38 by bone marrow cells in a cohort of patients with MM and indolent monoclonal gammopathies. Results indicate that only plasma cells expressed CD38 at high level within the bone niche. In addition, the flow cytometry analysis shows that CD38 was also expressed by monocytes and early osteoclast progenitors but not by osteoblasts and mature osteoclasts. Indeed, CD38 was lost during
Sigma-Aldrich
Anti-CD38 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.