MilliporeSigma
  • Methionine Sulfoxide Reductase A (MsrA) and Its Function in Ubiquitin-Like Protein Modification in
Methionine Sulfoxide Reductase A (MsrA) and Its Function in Ubiquitin-Like Protein Modification in
mBio (2017-09-07)
Xian Fu, Zachary Adams, Rui Liu, Nathaniel L Hepowit, Yifei Wu, Connor F Bowmann, Jackob Moskovitz, Julie A Maupin-Furlow
ABSTRACT
Methionine sulfoxide reductase A (MsrA) is an antioxidant enzyme found in all domains of life that catalyzes the reduction of methionine-
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.