MilliporeSigma
  • Chromogenic substrates for horseshoe crab clotting enzyme. Its application for the assay of bacterial endotoxins.
Chromogenic substrates for horseshoe crab clotting enzyme. Its application for the assay of bacterial endotoxins.
Haemostasis (1978-01-01)
S Iwanaga, T Morita, T Harada, S Nakamura, M Niwa, K Takada, T Kimura, S Sakakibara
PMID658779
ABSTRACT
An endotoxin-activated hemocyte lysate from the horseshoe crab (Tachypleus and Limulus) was found to hydrolyze Bz-Ile-Glu-(gamma-OR)-Gly-Arg-p-nitroanilide (PNA), Bz-Val-Gly-Arg-PNA, Boc-Val-Leu-Gly-Arg-PNA, and Boc-Leu-Gly-Arg-PNA, all of which have the COOH-terminal Gly-Arg sequence. This amidase activity was due to a clottting enzyme contained in the lysate. Furthermore, the amidase activity increased by increasing the concentration of bacterial endotoxin (E. coli, 0111-B4) added to the lysate. Therefore, the measurement of the endotoxin-induced amidase activity made it possible to determine the concentration of the endotoxin.