MilliporeSigma
  • Cytotoxicity of L-cycloserine against human neuroblastoma and medulloblastoma cells is associated with the suppression of ganglioside expression.
Cytotoxicity of L-cycloserine against human neuroblastoma and medulloblastoma cells is associated with the suppression of ganglioside expression.
Anticancer research (2000-03-04)
J Cinatl, J Cinatl, R Kotchetkov, P Pouckova, J U Vogel, H Rabenau, M Michaelis, B Kornhuber
ABSTRACT
Human neuroblastoma and medulloblastoma express abnormal ganglioside patterns especially GD2 and GM2 which are important for tumour growth. We tested the effects of L-cycloserine (L-CS), a potent inhibitor of synthesis of glycosphingolipids, on the growth, viability and expression of GD2 and GM2 in neuroblastoma and medulloblastoma cells. The cytotoxic effects of L-CS were tested using the MTT dye reduction assay on four neuroblastoma (IMR-32, SK-N-SH, UKF-NB-2 and UKF-NB-3), two medulloblastoma (D283 and D341) and normal human fibroblasts and epithelial cell lines. In some experiments cytotoxicity of L-CS was tested in the presence of exogenous GD2 and GM2. The expression of GD2 and GM2 was analysed by flow cytometry. The antitumoral effects of L-CS in vivo were assessed on established xenografts of UKF-NB-3 or D283 cells in athymic (nude) mice using systemic administration of the drug (150 mg/kg intraperitoneally, once per day on 20 consecutive days). In vitro experiments revealed that L-CS was toxic for tumour cells at concentrations ranging from 0.5 to 20 micrograms/ml without any significant effects on normal fibroblasts and epithelial cells. L-CS treatment of UKF-NB-3 and D283 cells significantly inhibited expression of GD2 and GM2. The addition of exogenous GD2 and GM2 to culture medium partially prevented cytotoxic effects of L-CS on tumour cells. In vivo treatment resulted in complete tumour regression of UKF-NB-3 xenografts whereas growth of D283 xenografts was reduced by 60%. L-CS is a selective antitumoral agent for neuroblastoma and medulloblastoma cells with the ability to reduce expression of tumour associated gangliosides. In vivo experiments suggest that L-CS may be effective drug for treatment of neuroblastoma and medulloblastoma.
Sigma-Aldrich
L-Cycloserine
Pricing and availability is not currently available.