- Mildly Cross-Linked Dendrimer Hydrogel Prepared via Aza-Michael Addition Reaction for Topical Brimonidine Delivery.
Mildly Cross-Linked Dendrimer Hydrogel Prepared via Aza-Michael Addition Reaction for Topical Brimonidine Delivery.
Journal of biomedical nanotechnology (2018-02-27)
Juan Wang, Geoffrey S Williamson, Michael G Lancina, Hu Yang
PMID29479294
ABSTRACT
In this work, we developed a mildly cross-linked dendrimer hydrogel (mcDH) via