MilliporeSigma
  • General method for synthesis of 2-heterocyclic N-methyliminodiacetic acid boronates.
General method for synthesis of 2-heterocyclic N-methyliminodiacetic acid boronates.
Organic letters (2010-05-15)
Graham R Dick, David M Knapp, Eric P Gillis, Martin D Burke
ABSTRACT
A wide range of 2-pyridyl and other difficult-to-access heterocyclic N-methyliminodiacetic acid boronates can be readily prepared from the corresponding bromides via a new method involving direct transligation of 2-heterocyclic trialkoxyborate salts with N-methyliminodiacetic acid (MIDA) at elevated temperatures.