MilliporeSigma
  • IGFBP2 induces SPRY1 expression via NF-κB signaling pathway in glioblastoma multiforme (GBM).
IGFBP2 induces SPRY1 expression via NF-κB signaling pathway in glioblastoma multiforme (GBM).
European review for medical and pharmacological sciences (2017-12-12)
Z-S Yuan, Y Cao, Z-Y Li
ABSTRACT
Gliomas are accompanied with high mortality owning to their invasive peculiarity and vulnerability to drug resistance. miR-21 is a vital oncogenic miRNA that regulates drug resistance of tumor cells. This study aims to elucidate the function of miR-21 in human glioma cells resistant to carmustine (BCNU) and to demonstrate the underlying molecular mechanism. BCNU-sensitive cells (SWOZ2 cells) were transfected with miR-21 agomir and negative control, and BCNU-resistance cells (SWOZ2-BCNU cells) were transfected with miR-21 antagomir and negative control. The Real-time fluorescence quantitative PCR was used to detect and compare the levels of miR-21expression between SWOZ2-BCNU and SWOZ2 cells. The drug sensitivity of these cells to BCNU was determined by Cell Counting Kit-8 (CCK-8) assay. The protein expression of Spry2 was detected by Western blotting. The expression level of miR-21 was remarkably higher in SWOZ2-BCNU cells than that in SWOZ2 cells. The half-maximal inhibitory concentration (IC50) of BCNU was obviously higher for SWOZ2-BCNU cells than that for SWOZ2 cells. Besides, we found that aberrant expression of miR-21 in SWOZ2-BCNU cells is responsible for glioma BCNU-resistance. Consistently, Spry2 protein levels were significantly reduced in SWOZ2-BCNU as well as in miR-21 agomir-transfected cells, inversely correlated to miR-21 expression. The results of si-Spry2 co-transfection suggested that the effect of miR-21 on glioma BCNU-resistance is mediated through Spry2. miR-21 enhances the resistance of human glioma cells to BCNU by decreasing the expression of Spry2 protein. Thus, Spry2 may be a novel therapeutic target for treating glioma BCNU-resistance.
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP2
Sigma-Aldrich
MISSION® esiRNA, targeting human SPRY1