MilliporeSigma
  • Florfenicol pharmacokinetics following intravenous and oral administrations and its elimination after oral and bath administrations in common carp
Florfenicol pharmacokinetics following intravenous and oral administrations and its elimination after oral and bath administrations in common carp
Veterinary research forum : an international quarterly journal (2018-01-13)
Abdolhossein Jangaran Nejad, Rahim Peyghan, Hossein Najafzadeh Varzi, Ali Shahriyari
ABSTRACT
The aim of this study was to evaluate pharmacokinetic profiles of florfenicol after a single dose of intravenous (5.00 mg kg
Product Description
Supelco
Florfenicol, analytical standard, for drug analysis