MilliporeSigma
  • Cellular volume and marker expression in human peripheral blood apheresis stem cells.
Cellular volume and marker expression in human peripheral blood apheresis stem cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2008-01-15)
Siddharth Sharma, Raquel Cabana, Sherry Shariatmadar, Awtar Krishan
ABSTRACT
Coulter volume is far more accurate measure of cell volume than forward angle light scatter. In this report, we have used Coulter volume to determine the mean cell volume and diameter of normal human peripheral blood cells and hematopoietic progenitor cells obtained by apheresis (HPC-A) from patients with hematological malignancies. Fresh peripheral blood samples (treated with Beckman Coulter IMMUNOPrep erythrocyte lysis solution), HPC-A samples (treated with BD Biosciences FACSLysing solution), or processed by Ficoll Hypaque sedimentation method were stained with CD45-FITC and PE-labeled CD34, CD90, CD117, and CD133 antibodies and analyzed for electronic volume and two color fluorescence. The mean electronic volume and diameter of mononuclear cells from fresh peripheral blood samples prepared with IMMUNOPrep were lymphocytes (191 microm(3), 7.16 microm), monocytes (370 microm(3), 9.91 microm), and granulocytes (328 microm(3), 8.56 microm). In mononuclear cells of HPC-A samples prepared by Histopaque-1077 sedimentation, the lymphocytes had volume and diameter of 311 microm(3), 8.4 microm, monocytes were 486 microm(3), 9.76 microm, and granulocytes were 515 microm(3), 9.95 microm. In contrast, HPC-A samples prepared after lysis with FACSLysing solution had mean electronic volume and diameter of lymphocytes (414 microm(3), 9.25 microm), monocytes (797 microm(3), 11.5 microm), and granulocytes (670 microm(3), 10.85 microm). Cell volume of mononuclear cells in the HPC-A samples prepared by Histopaque-1077 sedimentation method was correlated with the expression of stem cell markers CD34, CD90, CD117, and CD133. CD90 positive cells had the smallest mean electronic volume of 299.93 microm(3) when compared with cells with positive expression of CD133 (322 microm(3)), CD117 (349 microm(3)), CD34 (407 microm(3)), and CD45 (453 microm(3)). Correlation of cell volume with stem cell marker expression may allow for the identification of small stem cells, which may not express the conventional markers used for the identification of stem cells in HPC-A samples.
Sigma-Aldrich
Histopaque®-1077 Hybri-Max, liquid, sterile-filtered, BioReagent, suitable for hybridoma