MilliporeSigma
  • Endothelial nitric oxide synthase and cyclooxygenase are activated by hydrogen peroxide in renal hypertensive rat aorta.
Endothelial nitric oxide synthase and cyclooxygenase are activated by hydrogen peroxide in renal hypertensive rat aorta.
European journal of pharmacology (2017-08-07)
B R Silva, L Pernomian, T D De Paula, M D Grando, L M Bendhack
ABSTRACT
In this work, we hypothesized that cyclooxygenase (COX) activity can be regulated by nitric oxide (NO) and hydrogen peroxide (H
Sigma-Aldrich
SC-236, ≥98% (HPLC)