MilliporeSigma
Structure-driven CO
Scientific reports (2017-12-10)
Hidenori Hashimoto, Tsutomu Yamaguchi, Hiroyuki Ozeki, Sanehiro Muromachi
ABSTRACT
Ionic clathrate hydrates can selectively capture small gas molecules such as CO
Sigma-Aldrich
TBPB, ≥98% (HPLC)