MilliporeSigma
  • Nutritional and Phytochemical Composition of Vaccinium padifolium Sm Wild Berries and Radical Scavenging Activity.
Nutritional and Phytochemical Composition of Vaccinium padifolium Sm Wild Berries and Radical Scavenging Activity.
Journal of food science (2017-09-30)
Maria J Carvalho, Carla S Gouveia, Ana C Vieira, Ana C Pereira, Miguel  Carvalho, José C Marques
ABSTRACT
Blueberries have a well-deserved reputation as a potential functional food, supported by studies which have identified and quantified various nutrients and bioactive phytochemicals with known benefits for human diet and health. Wild blueberries have attracted particular attention due to the levels and concentrations of those phytonutrients. This study aims to evaluate for the first time the chemical composition of Madeira Island's endemic Vaccinium padifolium Sm wild berry. Results show that this fruit contains high values of total soluble phenolic content (around 4 g GAE kg This study is in line with the efforts of the scientific community to identify new sources of phytonutrients that are beneficial to human health, characterizing the wild Madeira blueberry in terms of phytonutrients that suggest there may be health benefits associated with its consumption. The findings of this research are very important for both the commercial and agricultural sectors that produce this fruit, and for consumers who seek phytonutrient-rich foods.
Product Description
Supelco
Total Dietary Fiber Assay Kit, sufficient for ~100 assays