MilliporeSigma
  • Metabolome analysis reveals a role for glyceraldehyde 3-phosphate dehydrogenase in the inhibition of
Metabolome analysis reveals a role for glyceraldehyde 3-phosphate dehydrogenase in the inhibition of
Biotechnology for biofuels (2017-12-08)
Liang Tian, Skyler J Perot, David Stevenson, Tyler Jacobson, Anthony A Lanahan, Daniel Amador-Noguez, Daniel G Olson, Lee R Lynd
ABSTRACT
Here, we analyzed changes in the relative concentration of intracellular metabolites in response to gradual addition of ethanol to growing cultures. For We have demonstrated that a metabolic bottleneck occurs at the GAPDH reaction when the growth of
Product Description
Supelco
Terrific Broth, modified, powder