MilliporeSigma
  • Simultaneous removal of sulfides and benzene in FCC gasoline by in situ hydrogenation over NiLaIn/ZrO
Simultaneous removal of sulfides and benzene in FCC gasoline by in situ hydrogenation over NiLaIn/ZrO
Journal of hazardous materials (2017-09-20)
Lei Wang, Qian Liu, Chunyu Jing, Jiajia Yin, Nchare Mominou, Shuzhen Li
ABSTRACT
The ZrO
Sigma-Aldrich
2-Methylthianaphthene, 97%
Pricing and availability is not currently available.