MilliporeSigma
  • Comparison of predictability for human pharmacokinetics parameters among monkeys, rats, and chimeric mice with humanised liver.
Comparison of predictability for human pharmacokinetics parameters among monkeys, rats, and chimeric mice with humanised liver.
Xenobiotica; the fate of foreign compounds in biological systems (2016-11-29)
Maki Miyamoto, Shinji Iwasaki, Ikumi Chisaki, Sayaka Nakagawa, Nobuyuki Amano, Hideki Hirabayashi
ABSTRACT
1. The aim of the present study was to evaluate the usefulness of chimeric mice with humanised liver (PXB mice) for the prediction of clearance (CL
Sigma-Aldrich
Carbazeran, ≥95% (HPLC)