MilliporeSigma
  • Conjugation of siRNA with comb-type PEG enhances serum stability and gene silencing efficiency.
Conjugation of siRNA with comb-type PEG enhances serum stability and gene silencing efficiency.
Macromolecular rapid communications (2011-04-12)
Karthikeyan Gunasekaran, Thi H Nguyen, Heather D Maynard, Thomas P Davis, Volga Bulmus
ABSTRACT
A thiol-modified siRNA targeting the enhanced green fluorescence protein (eGFP) gene was conjugated with RAFT-synthesized, pyridyl disulfide-functional poly(PEG methyl ether acrylate)s (p(PEGA)s). siRNA-p(PEGA) conjugates demonstrated significantly enhanced in vitro serum stability and nuclease resistance compared to the unmodified and thiol-modified siRNA. The complexes of siRNA-p(PEGA) conjugates with a fusogenic peptide, KALA ((+)/(-) = 2) inhibited the protein expression approximately 28-fold more than the KALA complex of the unmodified siRNA. The protein inhibition caused by siRNA-p(PEGA)-KALA complexes (56 ± 5%-58 ± 3% of the fluorescence expressed in non-treated cells) was comparable to the effect of the unmodified siRNA-lipofectamine complex (77 ± 7%).
Sigma-Aldrich
Poly(M(EO)2MA:Poly(OEGMA) 90:10, hydrazide functionalized, 25 wt. % (solution in water)
Sigma-Aldrich
Poly(OEGMA), hydrazide functionalized, 25 wt% solution in water
Sigma-Aldrich
Poly(M(EO)2MA:Poly(OEGMA), (90:10), aldehyde functionalized, 25 wt% solution in water