MilliporeSigma
  • Effect of Nitrate Leaching Caused by Swine Manure Application in Fields of the Yellow River Irrigation Zone of Ningxia, China.
Effect of Nitrate Leaching Caused by Swine Manure Application in Fields of the Yellow River Irrigation Zone of Ningxia, China.
Scientific reports (2017-10-24)
Shiqi Yang, Yongsheng Wang, Ruliang Liu, Aiping Zhang, Zhengli Yang
ABSTRACT
A five-year swine manure application trial and a study of nitrate leaching losses have been conducted. There were three treatments: traditional without manure (CK), traditional matched manure 4500 kg ha