MilliporeSigma
  • The fluorescently responsive 3-(naphthalen-1-ylethynyl)-3-deaza-2'-deoxyguanosine discriminates cytidine via the DNA minor groove.
The fluorescently responsive 3-(naphthalen-1-ylethynyl)-3-deaza-2'-deoxyguanosine discriminates cytidine via the DNA minor groove.
Organic & biomolecular chemistry (2017-09-08)
Azusa Suzuki, Masaki Yanagi, Takuya Takeda, Robert H E Hudson, Yoshio Saito
ABSTRACT
A new environmentally responsive fluorescent nucleoside, 3-(naphthalen-1-ylethynyl)-3-deaza-2'-deoxyguanosine (