- Automated synthesis of 1-[
Automated synthesis of 1-[
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2017-08-15)
Kiran Kumar Solingapuram Sai, H Donald Gage, Frankis Almaguel, Bryan Neth, Timothy M Hughes, Sebastien Tremblay, Christian-Alexandre Castellano, Stephen C Cunnane, Matthew J Jorgensen, Suzanne Craft, Akiva Mintz
PMID28806598
ABSTRACT
We automated radiochemical synthesis of 1-[