MilliporeSigma
  • Solid-Phase Synthesis of Difficult Purine-Rich PNAs through Selective Hmb Incorporation: Application to the Total Synthesis of Cell Penetrating Peptide-PNAs.
Solid-Phase Synthesis of Difficult Purine-Rich PNAs through Selective Hmb Incorporation: Application to the Total Synthesis of Cell Penetrating Peptide-PNAs.
Frontiers in chemistry (2017-11-03)
Julien Tailhades, Hotake Takizawa, Michael J Gait, Don A Wellings, John D Wade, Yoshitsugu Aoki, Fazel Shabanpoor
ABSTRACT
Antisense oligonucleotide (ASO)-based drug development is gaining significant momentum following the recent FDA approval of Eteplirsen (an ASO based on phosphorodiamidate morpholino) and Spinraza (2'-
Sigma-Aldrich
2,4-Dimethoxybenzaldehyde, 98%
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%