MilliporeSigma
Phlorotannin Composition of Laminaria digitata.
Phytochemical analysis : PCA (2017-06-15)
Anne M Vissers, Augusta Caligiani, Stefano Sforza, Jean-Paul Vincken, Harry Gruppen
ABSTRACT
Phlorotannins are complex mixtures of phloroglucinol oligomers connected via C-C (fucols) or C-O-C (phlorethols) linkages. Their uniformity in subunits and large molecular weight hamper their structural analysis. Despite its commercial relevance for alginate extraction, phlorotannins in Laminaria digitata have not been studied. To obtain quantitative and structural information on phlorotannins in a methanolic extract from L. digitata. The combined use of The fucol-to-phlorethol linkage ratio was 1:26 and the extract was 60.1% pure, determined by NMR spectroscopy. For DMBA, the response of the extract was 12 times lower than that of phloroglucinol, whereas there was no difference for FC. By accounting for differences in response, the colorimetric assays were applicable for quantification using phloroglucinol as a standard. The phlorotannin content was around 4.5% DM. Fucol- and phlorethol-linkage types were annotated based on characteristic MS By combining several analytical techniques, phlorotannins in L. digitata were quantified and characterised with respect to fucol-to-phlorethol linkage ratio, molecular weight (distribution), and occurrence of structural isomers. Copyright © 2017 John Wiley & Sons, Ltd.
Sigma-Aldrich
2,4-Dimethoxybenzaldehyde, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Pricing and availability is not currently available.