MilliporeSigma
High Manganese Tolerance and Biooxidation Ability of
Frontiers in microbiology (2017-10-25)
Natália R Barboza, Mônica M C A Morais, Pollyana S Queiroz, Soraya S Amorim, Renata Guerra-Sá, Versiane A Leão
ABSTRACT
Manganese is an important metal for the maintenance of several biological functions, but it can be toxic in high concentrations. One of the main forms of human exposure to metals, such as manganese (Mn), is the consumption of solar salt contaminated. Mn-tolerant bacteria could be used to decrease the concentration of this metal from contaminated sites through safer environmental-friendly alternative technology in the future. Therefore, this study was undertaken to isolate and identify Mn resistant bacteria from water samples collected from a Mn mine in the Iron Quadrangle region (Minas Gerais, Brazil). Two bacterial isolates were identified as
Sigma-Aldrich
Leucoberbelin Blue I, Dye content 65 %