MilliporeSigma
  • Multi-class analysis for simultaneous determination of pesticides, mycotoxins, process-induced toxicants and packaging contaminants in tea.
Multi-class analysis for simultaneous determination of pesticides, mycotoxins, process-induced toxicants and packaging contaminants in tea.
Food chemistry (2017-10-19)
Mathieu Cladière, Grégoire Delaporte, Even Le Roux, Valérie Camel
ABSTRACT
This study attempts at uniting the analysis of four different classes of contaminants for both liquid and solid tea samples. A total of 32 compounds, classified as pesticides, mycotoxins, process-induced toxicants or packaging contaminants, were carefully chosen for their diversity of structures and physicochemical properties. The proposed method combines a sample treatment strategy coming from metabolomics with liquid chromatography analysis using a silica bonded C18-pentafluorophenyl column coupled to high resolution mass spectrometry. For tea brew, dilute and shoot method provides good quantification (70-120% recoveries and <20% RSD) for more than 80% of compounds. For tea leaves, strong matrix effects are observed, thus, matrix-matched calibration is required to reach good performances, i.e. 63% of compounds quantified and 81% detected at 10µg/kg. Finally, method performances were evaluated against existing regulations, and it appears that 69% of contaminants are quantified and 91% detected at levels lower than their respective European regulation limits.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Supelco
Dinoseb, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland