MilliporeSigma
Magnetic Properties of CoFe
ACS applied materials & interfaces (2017-09-20)
Calvin D Pham, Jeffrey Chang, Mark A Zurbuchen, Jane P Chang
ABSTRACT
A radical-enhanced atomic layer deposition (RE-ALD) process was developed for growing ferrimagnetic CoFe