MilliporeSigma
DOCK8 Deficiency Presenting as an IPEX-Like Disorder.
Journal of clinical immunology (2017-10-24)
Fayhan J Alroqi, Louis-Marie Charbonnier, Sevgi Keles, Fatima Ghandour, Pierre Mouawad, Rami Sabouneh, Reem Mohammed, Abduarahman Almutairi, Janet Chou, Michel J Massaad, Raif S Geha, Zeina Baz, Talal A Chatila
ABSTRACT
The dedicator of cytokinesis 8 (DOCK8) deficiency is an autosomal recessive-combined immunodeficiency whose clinical spectra include recurrent infections, autoimmunity, malignancies, elevated serum IgE, eczema, and food allergies. Here, we report on patients with loss of function DOCK8 mutations with profound immune dysregulation suggestive of an immune dysregulation, polyendocrinopathy, enteropathy, X-linked (IPEX)-like disorder. Immunophenotyping of lymphocyte subpopulations and analysis of DOCK8 protein expression were evaluated by flow cytometry. T regulatory (T Patient 1 (P1) presented at 10 months of age with chronic severe diarrhea and active colitis in the absence of an infectious trigger, severe eczema with elevated serum IgE, and autoimmune hemolytic anemia, suggestive of an IPEX-related disorder. Whole-exome sequencing revealed a homozygous nonsense mutation in DOCK8 at the DOCK-homology region (DHR)-1 (c.1498C>T; p. R500X). Patient P2, a cousin of P1 who carries the same DOCK8 nonsense mutation, presented with eczema and recurrent ear infections in early infancy, and she developed persistent diarrhea by 3 years of age. Patient P3 presented with lymphoproliferation, severe eczema with allergic dysregulation, and chronic diarrhea with colitis. She harbored a homozygous loss of function DOCK8 mutation (c.2402 -1G→A). T DOCK8 deficiency may present severe immune dysregulation with features that may overlap with those of IPEX and other IPEX-like disorders.