MilliporeSigma
  • Simulating Bilayers of Nonionic Surfactants with the GROMOS-Compatible 2016H66 Force Field.
Simulating Bilayers of Nonionic Surfactants with the GROMOS-Compatible 2016H66 Force Field.
Langmuir : the ACS journal of surfaces and colloids (2017-08-24)
Caroline Senac, Wladimir Urbach, Erol Kurtisovski, Philippe H Hünenberger, Bruno A C Horta, Nicolas Taulier, Patrick F J Fuchs
ABSTRACT
Polyoxyethylene glycol alkyl ether amphiphiles (C
Sigma-Aldrich
Tetraethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)