MilliporeSigma
Site preferences in hetero-metallic [Fe
Dalton transactions (Cambridge, England : 2003) (2017-09-19)
Anastasia N Georgopoulou, Karrar Al-Ameed, Athanassios K Boudalis, Dimitrios F Anagnostopoulos, Vassilis Psycharis, John E McGrady, Yiannis Sanakis, Catherine P Raptopoulou
ABSTRACT
The reaction of mixtures of Fe(O
Sigma-Aldrich
Di(2-pyridyl) ketone, 99%