MilliporeSigma
  • Achieving Stable Mainstream Nitrogen Removal via the Nitrite Pathway by Sludge Treatment Using Free Ammonia.
Achieving Stable Mainstream Nitrogen Removal via the Nitrite Pathway by Sludge Treatment Using Free Ammonia.
Environmental science & technology (2017-08-05)
Qilin Wang, Haoran Duan, Wei Wei, Bing-Jie Ni, Andrew Laloo, Zhiguo Yuan
ABSTRACT
Biological nitrogen removal through the nitrite pathway (NH