MilliporeSigma
Physiological growth and galactose utilization by dairy yeast
3 Biotech (2017-09-29)
Arun Beniwal, Priyanka Saini, Anusha Kokkiligadda, Shilpa Vij
ABSTRACT
The dairy yeast
Sigma-Aldrich
β-Galactosidase from Kluyveromyces lactis, ≥2600 units/g