MilliporeSigma
  • Compound-specific isotope analysis resolves the dietary origin of docosahexaenoic acid in the mouse brain.
Compound-specific isotope analysis resolves the dietary origin of docosahexaenoic acid in the mouse brain.
Journal of lipid research (2017-07-12)
R J Scott Lacombe, Vanessa Giuliano, Stefanie M Colombo, Michael T Arts, Richard P Bazinet
ABSTRACT
DHA (22:6n-3) may be derived from two dietary sources, preformed dietary DHA or through synthesis from α-linolenic acid (ALA; 18:3n-3). However, conventional methods cannot distinguish between DHA derived from either source without the use of costly labeled tracers. In the present study, we demonstrate the proof-of-concept that compound-specific isotope analysis (CSIA) by GC-isotope ratio mass spectrometry (IRMS) can differentiate between sources of brain DHA based on differences in natural
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Pricing and availability is not currently available.