- The Impacts of Genistein and Daidzein on Estrogen Conjugations in Human Breast Cancer Cells: A Targeted Metabolomics Approach.
The Impacts of Genistein and Daidzein on Estrogen Conjugations in Human Breast Cancer Cells: A Targeted Metabolomics Approach.
Frontiers in pharmacology (2017-10-21)
Stefan Poschner, Alexandra Maier-Salamon, Martin Zehl, Judith Wackerlig, Daniel Dobusch, Bettina Pachmann, Konstantin L Sterlini, Walter Jäger
PMID29051735
ABSTRACT
The beneficial effect of dietary soy food intake, especially for women diagnosed with breast cancer, is controversial, as