MilliporeSigma
Unveiling a Drift Resistant Cryptotope within
Frontiers in immunology (2017-10-19)
John Anthony Garza, Alexander Bryan Taylor, Laura Jo Sherwood, Peter John Hart, Andrew Hayhurst
ABSTRACT
Marburg virus (MARV) is a highly lethal hemorrhagic fever virus that is increasingly re-emerging in Africa, has been imported to both Europe and the US, and is also a Tier 1 bioterror threat. As a negative sense RNA virus, MARV has error prone replication which can yield progeny capable of evading countermeasures. To evaluate this vulnerability, we sought to determine the epitopes of 4 llama single-domain antibodies (sdAbs or VHH) specific for nucleoprotein (NP), each capable of forming MARV monoclonal affinity reagent sandwich assays. Here, we show that all sdAb bound the C-terminal region of NP, which was produced recombinantly to derive X-ray crystal structures of the three best performing antibody-antigen complexes. The common epitope is a trio of alpha helices that form a novel asymmetric basin-like depression that accommodates each sdAb paratope
Sigma-Aldrich
Sodium oxamate, ≥98%