MilliporeSigma
  • Mechanistic Insight Leads to a Ligand Which Facilitates the Palladium-Catalyzed Formation of 2-(Hetero)Arylaminooxazoles and 4-(Hetero)Arylaminothiazoles.
Mechanistic Insight Leads to a Ligand Which Facilitates the Palladium-Catalyzed Formation of 2-(Hetero)Arylaminooxazoles and 4-(Hetero)Arylaminothiazoles.
Angewandte Chemie (International ed. in English) (2017-06-13)
Esben P K Olsen, Pedro L Arrechea, Stephen L Buchwald
ABSTRACT
By using mechanistic insight, a new ligand (EPhos) for the palladium-catalyzed C-N cross-coupling between primary amines and aryl halides has been developed. Employing an isopropoxy group at the C3-position favors the C-bound isomer of the ligand-supported palladium(II) complexes and leads to significantly improved reactivity. The use of a catalyst system based on EPhos with NaOPh as a mild homogeneous base proved to be very effective in the formation of 4-arylaminothiazoles and highly functionalized 2-arylaminooxazoles. Previously, these were not readily accessible using palladium catalysis.