MilliporeSigma
  • Altenusin, a Nonsteroidal Microbial Metabolite, Attenuates Nonalcoholic Fatty Liver Disease by Activating the Farnesoid X Receptor.
Altenusin, a Nonsteroidal Microbial Metabolite, Attenuates Nonalcoholic Fatty Liver Disease by Activating the Farnesoid X Receptor.
Molecular pharmacology (2017-07-26)
Zhihui Zheng, Zanmei Zhao, Shuqiang Li, Xinhua Lu, Mengxi Jiang, Jie Lin, Yunqi An, Yang Xie, Meishu Xu, Wenbin Shen, Grace L Guo, Yixian Huang, Song Li, Xuexia Zhang, Wen Xie
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is a prevalent chronic liver disease. The incidence of NAFLD has increased steadily due to its close association with the global epidemic of obesity and type 2 diabetes. However, there is no effective pharmacological therapy approved for NAFLD. Farnesoid X receptor (FXR), a member of the nuclear receptor subfamily, plays important roles in maintaining the homeostasis of bile acids, glucose, and lipids. FXR agonists have shown promise for the treatment of NAFLD. In this study, we report altenusin (2076A), a natural nonsteroidal fungal metabolite, as a novel selective agonist of FXR with an EC
Sigma-Aldrich
Altenusin, ≥98% (HPLC)