MilliporeSigma
  • Transformation of Unsaturated Fatty Acids/Esters to Corresponding Keto Fatty Acids/Esters by Aerobic Oxidation with Pd(II)/Lewis Acid Catalyst.
Transformation of Unsaturated Fatty Acids/Esters to Corresponding Keto Fatty Acids/Esters by Aerobic Oxidation with Pd(II)/Lewis Acid Catalyst.
Journal of agricultural and food chemistry (2017-07-19)
Ahmed M Senan, Sicheng Zhang, Miao Zeng, Zhuqi Chen, Guochuan Yin
ABSTRACT
Utilization of renewable biomass to partly replace the fossil resources in industrial applications has attracted attention due to the limited fossil feedstock with the increased environmental concerns. This work introduced a modified Wacker-type oxidation for transformation of unsaturated fatty acids/esters to the corresponding keto fatty acids/esters, in which Cu
Sigma-Aldrich
Copper(II) trifluoromethanesulfonate, 98%