MilliporeSigma
β-Catenin Directs Nuclear Factor-κB p65 Output
Frontiers in immunology (2017-09-26)
Tim Koopmans, Roos Eilers, Mark Menzen, Andrew Halayko, Reinoud Gosens
ABSTRACT
β-Catenin is a multifunctional protein that apart from its role in proliferative and differentiation events, also acts upon inflammatory processes, mainly
Sigma-Aldrich
MISSION® esiRNA, targeting human RELA