MilliporeSigma
  • Photoautotrophic production of polyhydroxyalkanoates in a synthetic mixed culture of
Photoautotrophic production of polyhydroxyalkanoates in a synthetic mixed culture of
Biotechnology for biofuels (2017-08-18)
Hannes Löwe, Karina Hobmeier, Manuel Moos, Andreas Kremling, Katharina Pflüger-Grau
ABSTRACT
One of the major challenges for the present and future generations is to find suitable substitutes for the fossil resources we rely on today. Cyanobacterial carbohydrates have been discussed as an emerging renewable feedstock in industrial biotechnology for the production of fuels and chemicals, showing promising production rates when compared to crop-based feedstock. However, intrinsic capacities of cyanobacteria to produce biotechnological compounds are limited and yields are low. Here, we present an approach to circumvent these problems by employing a synthetic bacterial co-culture for the carbon-neutral production of polyhydroxyalkanoates (PHAs) from CO These results demonstrate the feasibility of mixed cultures of
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%