MilliporeSigma
  • 1-Alkyl-3-methylimidazolium alkanesulfonate ionic liquids, [C(n)H(2)(n)(+1)mim][C(k)H(2)(k)(+1)SO(3)]: synthesis and physicochemical properties.
1-Alkyl-3-methylimidazolium alkanesulfonate ionic liquids, [C(n)H(2)(n)(+1)mim][C(k)H(2)(k)(+1)SO(3)]: synthesis and physicochemical properties.
Physical chemistry chemical physics : PCCP (2010-05-08)
Marijana Blesic, Małgorzata Swadźba-Kwaśny, Tayeb Belhocine, H Q Nimal Gunaratne, José N Canongia Lopes, Margarida F Costa Gomes, Agílio A H Pádua, Kenneth R Seddon, Luís Paulo N Rebelo
ABSTRACT
A set of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids, [C(n)mim][C(k)SO(3)], formed by the variation of the alkyl chain lengths both in the cation and the anion (n = 1-6, 8, or 10; k = 1-4, or 6), was synthesised, with sixteen of them being novel. The ionic liquids were characterised by (1)H and (13)C NMR spectroscopy, and mass spectrometry. Their viscosities and densities as a function of temperature, as well as melting points and decomposition temperatures, were determined. The molecular volumes, both experimental and calculated, were found to depend linearly on the sum (n + k).
Sigma-Aldrich
1-Propanesulfonyl chloride, 97%
Pricing and availability is not currently available.