MilliporeSigma
Strong tissue glue with tunable elasticity.
Acta biomaterialia (2017-02-13)
Regina Kelmansky, Brian J McAlvin, Abraham Nyska, Jenny C Dohlman, Homer H Chiang, Michinao Hashimoto, Daniel S Kohane, Boaz Mizrahi
ABSTRACT
Many bio-adhesive materials adhere weakly to tissue due to their high water content and weak structural integrity. Others provide desirable adhesive strength but suffer from rigid structure and lack of elasticity after administration. We have developed two water-free, liquid four-armed PEG pre-polymers modified with NHS or with NH Many bio-adhesive materials adhere weakly to tissue (e.g. hydrogels) due to their high water content and weak structural integrity. Others provide desirable mechanical properties but suffer from poor biocompatibility (e.g. cyanoacrylates). This study proposes a new concept for the formation of super strong and tunable tissue glues. Our bio-materials' enhanced performance is the product of new neat (without water or other solvents) liquid polymers that solidify after administration while allowing interactions with the tissue. Moreover, the elastic modulus of these materials could easily be tuned without compromising biocompatibility. This system could be an attractive alternative to sutures and staples since it can be applied more quickly, causes less pain and may require less equipment while maintaining the desired adhesion strength.
Sigma-Aldrich
Diisopropyl azodicarboxylate, 98%
Pricing and availability is not currently available.