MilliporeSigma
  • An optimised clearing protocol for the quantitative assessment of sub-epidermal ovule tissues within whole cereal pistils.
An optimised clearing protocol for the quantitative assessment of sub-epidermal ovule tissues within whole cereal pistils.
Plant methods (2017-08-22)
Laura G Wilkinson, Matthew R Tucker
ABSTRACT
Seed development in the angiosperms requires the production of a female gametophyte (embryo sac) within the ovule. Many aspects of female reproductive development in cereal crops are yet to be described, largely due to the technical difficulty in obtaining phenotypic information at the cellular or sub-cellular level. Hoyer's solution is currently well established as a solution for clearing thin tissues samples, such as sections or whole tissues of bryophytes, mycorrhizal fungi, and small model organisms (e.g. Here we report a Hoyer's solution-based clearing method to facilitate clearing of the whole barley pistil, with high reproducibility. The clearing process takes 10 days from fixation to visualisation, whereupon tissue is sufficiently clear to obtain multiple phenotypic measurements from sub-epidermal tissues and cells within the ovule. Visualisation of cereal ovules that have not been dissected from the pistil allows an unprecedented capability to collect quantitative morphological information from the developing ovule, integument, nucellus and embryo sac. This will enable comparisons with genetic data to reveal the contribution of pre-fertilisation ovule tissues towards downstream seed development.
Product Description
Supelco
Liquid scintillation vials (with screw cap attached), white polypropylene cap, metal foil liner, pkg of 500 ea