MilliporeSigma
  • Manipulating PML SUMOylation via Silencing UBC9 and RNF4 Regulates Cardiac Fibrosis.
Manipulating PML SUMOylation via Silencing UBC9 and RNF4 Regulates Cardiac Fibrosis.
Molecular therapy : the journal of the American Society of Gene Therapy (2017-02-02)
Yu Liu, Dan Zhao, Fang Qiu, Ling-Ling Zhang, Shang-Kun Liu, Yuan-Yuan Li, Mei-Tong Liu, Di Wu, Jia-Xin Wang, Xiao-Qing Ding, Yan-Xin Liu, Chang-Jiang Dong, Xiao-Qi Shao, Bao-Feng Yang, Wen-Feng Chu
ABSTRACT
The promyelocytic leukemia protein (PML) is essential in the assembly of dynamic subnuclear structures called PML nuclear bodies (PML-NBs), which are involved in regulating diverse cellular functions. However, the possibility of PML being involved in cardiac disease has not been examined. In mice undergoing transverse aortic constriction (TAC) and arsenic trioxide (ATO) injection, transforming growth factor β1 (TGF-β1) was upregulated along with dynamic alteration of PML SUMOylation. In cultured neonatal mouse cardiac fibroblasts (NMCFs), ATO, angiotensin II (Ang II), and fetal bovine serum (FBS) significantly triggered PML SUMOylation and the assembly of PML-NBs. Inhibition of SUMOylated PML by silencing UBC9, the unique SUMO E2-conjugating enzyme, reduced the development of cardiac fibrosis and partially improved cardiac function in TAC mice. In contrast, enhancing SUMOylated PML accumulation, by silencing RNF4, a poly-SUMO-specific E3 ubiquitin ligase, accelerated the induction of cardiac fibrosis and promoted cardiac function injury. PML colocalized with Pin1 (a positive regulator for TGF-β1 mRNA expression in PML-NBs) and increased TGF-β1 activity. These findings suggest that the UBC9/PML/RNF4 axis plays a critical role as an important SUMO pathway in cardiac fibrosis. Modulating the protein levels of the pathway provides an attractive therapeutic target for the treatment of cardiac fibrosis and heart failure.
Sigma-Aldrich
Trichrome Stain (Masson) Kit
Pricing and availability is not currently available.