MilliporeSigma
  • Elevated auxin biosynthesis and transport underlie high vein density in C
Elevated auxin biosynthesis and transport underlie high vein density in C
Proceedings of the National Academy of Sciences of the United States of America (2017-08-02)
Chi-Fa Huang, Chun-Ping Yu, Yeh-Hua Wu, Mei-Yeh Jade Lu, Shih-Long Tu, Shu-Hsing Wu, Shin-Han Shiu, Maurice S B Ku, Wen-Hsiung Li
ABSTRACT
High vein density, a distinctive trait of C
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Indole-2,4,5,6,7-d5-3-acetic acid, 98 atom % D, 98% (CP)